Assay Details
Target Gene Details
Entrez Gene ID: | 23092 |
Gene Name: | Rho GTPase activating protein 26 |
Gene Aliases: |
GRAF, GRAF1, OPHN1L, OPHN1L1 |
Location: |
Chr.5:142770370-143229007 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP26 | NM_001135608.1 | NP_001129080.1 | ||
| NM_015071.4 | NP_055886.1 | |||
| XM_005268398.4 | XP_005268455.1 | |||
| XM_005268399.4 | XP_005268456.1 | |||
| XM_005268400.4 | XP_005268457.1 | |||
| XM_005268402.4 | XP_005268459.1 | |||
| XM_006714774.3 | XP_006714837.1 | |||
| XM_011537610.2 | XP_011535912.1 | |||
| XM_011537611.2 | XP_011535913.1 | |||
| XM_011537612.2 | XP_011535914.1 | |||
| XM_017009247.1 | XP_016864736.1 | |||
| XM_017009248.1 | XP_016864737.1 | |||
| XM_017009249.1 | XP_016864738.1 | |||
| XM_017009250.1 | XP_016864739.1 | |||
| BC068555.1 | AAH68555.1 | |||
| BF085236.1 | ||||
| KU178561.1 | ||||
| KU178562.1 |
Target Gene Details
Entrez Gene ID: | 100874239 |
Gene Name: | ARHGAP26 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:142859604-142868910 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP26-AS1 | NR_046680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606999 | Chr.5:142429551 - 143065624 on Build GRCh38 | Gain |
|
| nsv1031126 | Chr.5:142514198 - 143020006 on Build GRCh38 | Gain |
|
| esv3607011 | Chr.5:142795354 - 142880786 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map