Assay Details
Target Gene Details
Entrez Gene ID: | 55568 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 10 |
Gene Aliases: |
GALNACT10, PPGALNACT10, PPGANTASE10 |
Location: |
Chr.5:154190735-154420984 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT10 | NM_198321.3 | NP_938080.1 | ||
| AB078145.1 | BAC56890.1 | |||
| AJ505950.1 | CAD44532.1 | |||
| AK023782.1 | BAB14676.1 | |||
| AK127135.1 | ||||
| AL096739.1 | CAB46378.1 | |||
| BC007224.1 | AAH07224.2 | |||
| BC072450.1 | AAH72450.1 | |||
| BC098444.1 | AAH98444.1 |
Target Gene Details
Entrez Gene ID: | 386627 |
Gene Name: | SAP30L antisense RNA 1 (head to head) |
Gene Aliases: |
GALNT10-AS1 |
Location: |
Chr.5:154389769-154445822 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAP30L-AS1 | NR_037897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830520 | Chr.5:154372285 - 154553755 on Build GRCh38 | Loss |
|
| nsv1015362 | Chr.5:154215521 - 154450098 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map