Assay Details
Target Gene Details
Entrez Gene ID: | 10087 |
Gene Name: | collagen type IV alpha 3 binding protein |
Gene Aliases: |
CERT, CERTL, GPBP, MRD34, STARD11 |
Location: |
Chr.5:75368486-75511981 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL4A3BP | XM_006714513.2 | XP_006714576.1 | ||
| XM_017008919.1 | XP_016864408.1 | |||
| AK125387.1 | 1 | 1758 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575856 | Chr.5:75260838 - 75400813 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map