Assay Details
Target Gene Details
Entrez Gene ID: | 51128 |
Gene Name: | secretion associated Ras related GTPase 1B |
Gene Aliases: |
ANDD, CMRD, GTBPB, SARA2 |
Location: |
Chr.5:134601149-134632843 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAR1B | NM_001033503.2 | 8 | 3168 | NP_001028675.1 |
| NM_016103.3 | 7 | 3049 | NP_057187.1 | |
| XM_017009521.1 | 7 | 3056 | XP_016865010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520381 | Chr.5:134523918 - 134643239 on Build GRCh38 | Gain+Loss |
|
| nsv830491 | Chr.5:134557253 - 134724800 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77677325] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map