Assay Details
Target Gene Details
Entrez Gene ID: | 170591 |
Gene Name: | S100 calcium binding protein Z |
Gene Aliases: |
Gm625, S100-zeta |
Location: |
Chr.5:76849953-76952882 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100Z | NM_130772.3 | NP_570128.2 | ||
| XM_011543239.2 | XP_011541541.1 | |||
| XM_011543240.2 | XP_011541542.1 | |||
| XM_011543241.2 | XP_011541543.1 | |||
| XM_011543244.2 | XP_011541546.1 | |||
| XM_017009172.1 | XP_016864661.1 | |||
| XM_017009173.1 | XP_016864662.1 | |||
| BC022320.1 | AAH22320.1 | |||
| BX281742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34133 | Chr.5:76842871 - 76943505 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map