Assay Details
Target Gene Details
Entrez Gene ID: | 57486 |
Gene Name: | neurolysin |
Gene Aliases: |
AGTBP, EP24.16, MEP, MOP |
Location: |
Chr.5:65722196-65829283 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLN | NM_020726.4 | NP_065777.1 | ||
| XM_005248559.2 | XP_005248616.1 | |||
| XM_006714661.2 | XP_006714724.1 | |||
| XM_017009673.1 | XP_016865162.1 | |||
| AB033052.2 | BAA86540.2 | |||
| AJ300837.1 | CAC27329.1 | |||
| AK096416.1 | ||||
| AK292803.1 | ||||
| BC001644.2 | AAH01644.1 | |||
| BC004985.1 | AAH04985.1 | |||
| CB992921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3566030 | Chr.5:65732186 - 65734700 on Build GRCh38 | Deletion |
|
| esv3605303 | Chr.5:65732242 - 65734544 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map