Assay Details
Target Gene Details
Entrez Gene ID: | 10412 |
Gene Name: | NSA2, ribosome biogenesis homolog |
Gene Aliases: |
CDK105, HCL-G1, HCLG1, HUSSY-29, HUSSY29, TINP1 |
Location: |
Chr.5:74765429-74776912 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSA2 | NM_001271665.1 | NP_001258594.1 | ||
| NM_014886.4 | NP_055701.1 | |||
| NR_073403.1 | ||||
| XM_011543098.1 | XP_011541400.1 | |||
| XM_017008952.1 | XP_016864441.1 | |||
| XM_017008953.1 | XP_016864442.1 | |||
| AF077615.1 | AAG43048.1 | |||
| AF372458.1 | AAK53761.1 | |||
| AJ012409.1 | CAA10008.1 | |||
| AK313784.1 | ||||
| BC005288.1 | AAH05288.1 | |||
| HY021659.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv968947 | Chr.5:74768651 - 74769458 on Build GRCh38 | Duplication |
|
| nsv598653 | Chr.5:74697056 - 74799717 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map