Assay Details
Target Gene Details
Entrez Gene ID: | 133584 |
Gene Name: | EGF like, fibronectin type III and laminin G domains |
Gene Aliases: |
AGRINL, AGRNL, PIKA |
Location: |
Chr.5:38258409-38465480 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGFLAM | NM_001205301.1 | NP_001192230.1 | ||
| NM_152403.3 | NP_689616.2 | |||
| AK096474.1 | BAC04800.1 | |||
| AK291602.1 | ||||
| BC063822.1 | AAH63822.1 | |||
| BX647551.1 | CAH56137.1 |
Target Gene Details
Entrez Gene ID: | 100852408 |
Gene Name: | EGFLAM antisense RNA 4 |
Gene Aliases: |
- |
Location: |
Chr.5:38282269-38290986 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGFLAM-AS4 | NR_046219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604766 | Chr.5:38221565 - 38446015 on Build GRCh38 | Gain |
|
| esv3604761 | Chr.5:38056637 - 38805862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map