Assay Details
Target Gene Details
Entrez Gene ID: | 56979 |
Gene Name: | PR domain 9 |
Gene Aliases: |
MEISETZ, MSBP3, PFM6, ZNF899 |
Location: |
Chr.5:23507609-23528597 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDM9 | NM_001310214.1 | NP_001297143.1 | ||
| NM_020227.3 | NP_064612.2 | |||
| AF275816.1 | AAF87242.1 | |||
| AK301776.1 | ||||
| DQ388610.1 | ABD47939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762506 | Chr.5:22419490 - 24131170 on Build GRCh38 | Gain |
|
| nsv597544 | Chr.5:23495211 - 23571434 on Build GRCh38 | Gain |
|
| nsv1028347 | Chr.5:23407127 - 23686843 on Build GRCh38 | Loss |
|
| nsv1017510 | Chr.5:23473388 - 23525136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map