Assay Details
Target Gene Details
Entrez Gene ID: | 51128 |
Gene Name: | secretion associated Ras related GTPase 1B |
Gene Aliases: |
ANDD, CMRD, GTBPB, SARA2 |
Location: |
Chr.5:134601149-134632843 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAR1B | NM_001033503.2 | NP_001028675.1 | ||
| NM_016103.3 | NP_057187.1 | |||
| XM_017009521.1 | XP_016865010.1 | |||
| AF087850.1 | AAP97161.1 | |||
| AF092130.1 | AAD40372.1 | |||
| AK223452.1 | BAD97172.1 | |||
| AL512710.1 | CAC21652.1 | |||
| AL710660.1 | ||||
| BC002847.2 | AAH02847.1 | |||
| BC093034.1 | AAH93034.1 | |||
| CD048852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520381 | Chr.5:134523918 - 134643239 on Build GRCh38 | Gain+Loss |
|
| nsv830491 | Chr.5:134557253 - 134724800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map