Assay Details
Target Gene Details
Entrez Gene ID: | 1839 |
Gene Name: | heparin binding EGF like growth factor |
Gene Aliases: |
DTR, DTS, DTSF, HEGFL |
Location: |
Chr.5:140332843-140346603 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HBEGF | NM_001945.2 | NP_001936.1 | ||
| AK222688.1 | BAD96408.1 | |||
| AK307310.1 | ||||
| AK313202.1 | ||||
| BC033097.1 | AAH33097.1 | |||
| M60278.1 | AAA35956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509090 | Chr.5:140311306 - 140421024 on Build GRCh38 | Insertion |
|
| nsv1020011 | Chr.5:140340815 - 140396254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map