Assay Details
Target Gene Details
Entrez Gene ID: | 3977 |
Gene Name: | leukemia inhibitory factor receptor alpha |
Gene Aliases: |
CD118, LIF-R, SJS2, STWS, SWS |
Location: |
Chr.5:38474963-38606290 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIFR | NM_002310.5 | NP_002301.1 | ||
| XM_017009462.1 | XP_016864951.1 | |||
| XM_017009463.1 | XP_016864952.1 | |||
| AK308158.1 | ||||
| AK309882.1 | ||||
| DA835819.1 | ||||
| DA857408.1 | ||||
| X61615.1 | CAA43805.1 |
Target Gene Details
Entrez Gene ID: | 100506495 |
Gene Name: | LIFR antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:38556786-38671216 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIFR-AS1 | NR_103553.1 | |||
| NR_103554.1 | ||||
| BG771462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604761 | Chr.5:38056637 - 38805862 on Build GRCh38 | Gain |
|
| dgv1594e212 | Chr.5:38574890 - 38586004 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map