Assay Details
Target Gene Details
Entrez Gene ID: | 4724 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit S4 |
Gene Aliases: |
AQDQ, CI-18, CI-18 kDa, CI-AQDQ |
Location: |
Chr.5:53560610-53683341 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFS4 | NM_001318051.1 | NP_001304980.1 | ||
| NM_002495.3 | NP_002486.1 | |||
| NR_134473.1 | ||||
| NR_134474.1 | ||||
| NR_134475.1 | ||||
| XM_017009491.1 | XP_016864980.1 | |||
| AF020351.1 | AAB87865.1 | |||
| AK307973.1 | ||||
| BC005270.1 | AAH05270.1 | |||
| BU145342.1 | ||||
| BU954271.1 | ||||
| CD692133.1 | ||||
| DA609203.1 | ||||
| DB489661.1 | ||||
| KU178226.1 | ||||
| KU178227.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5666n100 | Chr.5:53231175 - 54453241 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map