Assay Details
Target Gene Details
Entrez Gene ID: | 4012 |
Gene Name: | leucyl and cystinyl aminopeptidase |
Gene Aliases: |
CAP, IRAP, P-LAP, PLAP |
Location: |
Chr.5:96935642-97029411 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNPEP | NM_005575.2 | NP_005566.2 | ||
| AK096804.1 | 2 | 1343 | ||
| AK314226.1 | ||||
| D50810.1 | BAA09436.1 | |||
| U62768.1 | AAB66672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598963 | Chr.5:96749072 - 97027703 on Build GRCh38 | Gain |
|
| nsv598962 | Chr.5:96722187 - 97557460 on Build GRCh38 | Gain |
|
| nsv1021263 | Chr.5:96060753 - 97448132 on Build GRCh38 | Loss |
|
| nsv1028741 | Chr.5:96746697 - 97036461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map