Assay Details
Target Gene Details
Entrez Gene ID: | 3572 |
Gene Name: | interleukin 6 signal transducer |
Gene Aliases: |
CD130, CDW130, GP130, IL-6RB |
Location: |
Chr.5:55935095-55994993 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL6ST | NM_001190981.1 | 16 | 7980 | NP_001177910.1 |
| NM_002184.3 | 17 | 8163 | NP_002175.2 | |
| NM_175767.2 | 16 | 8080 | NP_786943.1 | |
| NR_120480.1 | 18 | 8209 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830307 | Chr.5:55867930 - 56048443 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79149215] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map