Assay Details
Target Gene Details
Entrez Gene ID: | 91942 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 2 |
Gene Aliases: |
B17.2L, MMTN, NDUFA12L, mimitin |
Location: |
Chr.5:60945129-61153037 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF2 | NM_174889.4 | NP_777549.1 | ||
| AB183433.1 | BAD91205.1 | |||
| AF087990.1 | ||||
| AK026051.1 | ||||
| AK291296.1 | ||||
| BC022869.1 | ||||
| BC033965.1 | AAH33965.1 | |||
| KU178834.1 | ||||
| KU178835.1 | ||||
| KU178836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518695 | Chr.5:60971048 - 61033797 on Build GRCh38 | Loss |
|
| nsv1019307 | Chr.5:60999523 - 61113250 on Build GRCh38 | Gain |
|
| dgv9810n54 | Chr.5:60820786 - 61017188 on Build GRCh38 | Loss |
|
| nsv520871 | Chr.5:60845418 - 61024330 on Build GRCh38 | Loss |
|
| nsv462199 | Chr.5:60833534 - 61033797 on Build GRCh38 | Gain |
|
| nsv598313 | Chr.5:60951988 - 61079085 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map