Assay Details
Target Gene Details
Entrez Gene ID: | 5295 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 1 |
Gene Aliases: |
AGM7, GRB1, IMD36, p85, p85-ALPHA |
Location: |
Chr.5:68215737-68301821 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R1 | NM_181523.2 | NP_852664.1 | ||
| XM_005248542.3 | XP_005248599.1 | |||
| XM_017009585.1 | XP_016865074.1 | |||
| BC094795.1 | AAH94795.1 | |||
| M61906.1 |
More Information
Set Membership: |
|

Genomic Map