Assay Details
Target Gene Details
Entrez Gene ID: | 133396 |
Gene Name: | interleukin 31 receptor A |
Gene Aliases: |
CRL, CRL3, GLM-R, GLMR, GPL, IL-31RA, PLCA2, PRO21384, hGLM-R |
Location: |
Chr.5:55840334-55922854 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL31RA | XM_011543142.2 | XP_011541444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830306 | Chr.5:55673052 - 55846654 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map