Assay Details
Target Gene Details
Entrez Gene ID: | 27125 |
Gene Name: | AF4/FMR2 family member 4 |
Gene Aliases: |
AF5Q31, CHOPS, MCEF |
Location: |
Chr.5:132875379-132963662 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFF4 | NM_014423.3 | NP_055238.1 | ||
| XM_005271963.4 | XP_005272020.1 | |||
| XM_006714587.3 | XP_006714650.1 | |||
| AB209547.1 | BAD92784.1 | |||
| AF197927.1 | AAF18981.1 | |||
| AF213987.2 | AAM00184.2 | |||
| AK055638.1 | ||||
| AK307407.1 | ||||
| BC025700.1 | AAH25700.1 | |||
| BC063007.1 | AAH63007.1 | |||
| BC100287.1 | AAI00288.1 | |||
| BC137226.1 | ||||
| DA934874.1 |
Target Gene Details
Entrez Gene ID: | 105379177 |
Gene Name: | uncharacterized LOC105379177 |
Gene Aliases: |
- |
Location: |
Chr.5:132958461-132961580 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379177 | XM_011543785.2 | 1 | 2784 | XP_011542087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830490 | Chr.5:132953258 - 133115177 on Build GRCh38 | Gain |
|
| nsv830489 | Chr.5:132815845 - 133000744 on Build GRCh38 | Loss |
|
| nsv1022039 | Chr.5:132914933 - 133045063 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map