Assay Details
Target Gene Details
Entrez Gene ID: | 55374 |
Gene Name: | transmembrane and coiled-coil domains 6 |
Gene Aliases: |
PRO1580 |
Location: |
Chr.5:140596529-140647411 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMCO6 | XM_011537665.2 | XP_011535967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462462 | Chr.5:140556654 - 140626201 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map