Assay Details
Target Gene Details
Entrez Gene ID: | 5145 |
Gene Name: | phosphodiesterase 6A |
Gene Aliases: |
CGPR-A, PDEA, RP43 |
Location: |
Chr.5:149857956-149944793 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE6A | NM_000440.2 | 22 | 4270 | NP_000431.2 |
| XM_011537650.2 | 19 | 3401 | XP_011535952.1 | |
| XM_011537651.2 | 16 | 3869 | XP_011535953.1 | |
| XM_011537652.1 | 17 | 3339 | XP_011535954.1 | |
| XM_011537653.2 | 16 | 4035 | XP_011535955.1 | |
| XM_011537654.2 | 16 | 4032 | XP_011535956.1 | |
| XM_017009572.1 | 20 | 4027 | XP_016865061.1 | |
| AB593141.1 | ||||
| BX537592.1 | 1 | 1430 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023343 | Chr.5:149851224 - 149888162 on Build GRCh38 | Gain |
|
| esv24664 | Chr.5:149854140 - 150050084 on Build GRCh38 | Gain+Loss |
|
| esv3890751 | Chr.5:149851956 - 149886716 on Build GRCh38 | Gain |
|
| dgv727n27 | Chr.5:149851167 - 149881249 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map