Assay Details
Target Gene Details
Entrez Gene ID: | 3567 |
Gene Name: | interleukin 5 |
Gene Aliases: |
EDF, IL-5, TRF |
Location: |
Chr.5:132539194-132556827 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL5 | XM_005271988.3 | XP_005272045.1 | ||
XM_011543373.2 | XP_011541675.1 | |||
XM_011543374.2 | XP_011541676.1 | |||
DQ655971.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
