Assay Details
Target Gene Details
Entrez Gene ID: | 2651 |
Gene Name: | glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group) |
Gene Aliases: |
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1 |
Location: |
Chr.6:10521283-10629368 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600988 | Chr.6:10501021 - 10614845 on Build GRCh38 | Loss |
|
| nsv966670 | Chr.6:10447438 - 10871656 on Build GRCh38 | Duplication |
|
| nsv600979 | Chr.6:10449025 - 10839526 on Build GRCh38 | Gain |
|
| nsv470800 | Chr.6:10538711 - 10863092 on Build GRCh38 | Gain |
|
| esv2759406 | Chr.6:10443564 - 10642899 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map