Assay Details
Target Gene Details
Entrez Gene ID: | 23493 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif 2 |
Gene Aliases: |
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32 |
Location: |
Chr.6:125747592-125762243 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEY2 | XM_017010629.1 | XP_016866118.1 | ||
| AK303890.1 |
More Information
Set Membership: |
|

Genomic Map