Assay Details
Target Gene Details
Entrez Gene ID: | 9247 |
Gene Name: | glial cells missing homolog 2 |
Gene Aliases: |
GCMB, hGCMb |
Location: |
Chr.6:10873223-10881865 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCM2 | NM_004752.3 | NP_004743.1 | ||
| AF079550.1 | AAC33792.1 | |||
| AF091149.1 | AAC98097.1 | |||
| BC069603.1 | AAH69603.1 | |||
| BC117316.1 | ||||
| BC117318.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519302 | Chr.6:10879763 - 10892266 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map