Assay Details
Target Gene Details
Entrez Gene ID: | 254394 |
Gene Name: | minichromosome maintenance 9 homologous recombination repair factor |
Gene Aliases: |
C6orf61, MCMDC1, ODG4, dJ329L24.1, dJ329L24.3 |
Location: |
Chr.6:118813449-118935162 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM9 | NM_017696.2 | NP_060166.2 | ||
| AK299076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161446 | Chr.6:118418441 - 118902959 on Build GRCh38 | Duplication |
|
| esv2677245 | Chr.6:118818180 - 118834905 on Build GRCh38 | Deletion |
|
| nsv5453 | Chr.6:118792655 - 118825738 on Build GRCh38 | Insertion |
|
| nsv509151 | Chr.6:118823819 - 118901881 on Build GRCh38 | Insertion |
|
| nsv1020463 | Chr.6:118271865 - 118825200 on Build GRCh38 | Gain |
|
| esv3610527 | Chr.6:118818233 - 118834600 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map