Assay Details
Target Gene Details
Entrez Gene ID: | 29940 |
Gene Name: | dermatan sulfate epimerase |
Gene Aliases: |
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2 |
Location: |
Chr.6:116254152-116441261 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSE | NM_001080976.2 | NP_001074445.1 | ||
| NM_001322937.1 | NP_001309866.1 | |||
| NM_001322938.1 | NP_001309867.1 | |||
| NM_001322940.1 | NP_001309869.1 | |||
| NR_136521.1 | ||||
| NR_136522.1 | ||||
| NR_136523.1 | ||||
| NR_136524.1 | ||||
| XM_017010795.1 | XP_016866284.1 | |||
| AK095123.1 | ||||
| AK293872.1 | ||||
| BC039245.1 | AAH39245.1 | |||
| BX110619.1 | ||||
| DC333659.1 | ||||
| HY012067.1 | ||||
| HY050986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576211 | Chr.6:116095308 - 116431057 on Build GRCh38 | Gain |
|
| nsv523972 | Chr.6:115958024 - 116809541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map