Assay Details
Target Gene Details
Entrez Gene ID: | 64065 |
Gene Name: | PERP, TP53 apoptosis effector |
Gene Aliases: |
KCP1, KRTCAP1, PIGPC1, THW, dJ496H19.1 |
Location: |
Chr.6:138088505-138107523 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PERP | NM_022121.4 | 3 | 2962 | NP_071404.2 |
More Information
Set Membership: |
|

Genomic Map