Assay Details
Target Gene Details
Entrez Gene ID: | 51557 |
Gene Name: | lengsin, lens protein with glutamine synthetase domain |
Gene Aliases: |
GLULD1, LGS |
Location: |
Chr.6:63275951-63567921 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGSN | XM_017010929.1 | 1 | 2206 | XP_016866418.1 |
Target Gene Details
Entrez Gene ID: | 7803 |
Gene Name: | protein tyrosine phosphatase type IVA, member 1 |
Gene Aliases: |
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1 |
Location: |
Chr.6:63528021-63583588 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTP4A1 | XM_017011270.1 | XP_016866759.1 | ||
| CR749458.1 | CAH18292.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs66984720] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map