Assay Details
Target Gene Details
Entrez Gene ID: | 221442 |
Gene Name: | adenylate cyclase 10, soluble pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41101034-41140835 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY10P1 | NR_026938.2 | |||
| BX648193.1 |
Target Gene Details
Entrez Gene ID: | 4800 |
Gene Name: | nuclear transcription factor Y subunit alpha |
Gene Aliases: |
CBF-A, CBF-B, HAP2, NF-YA |
Location: |
Chr.6:41072968-41102407 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYA | NM_002505.4 | 10 | 5920 | NP_002496.1 |
| NM_021705.3 | 9 | 5833 | NP_068351.1 | |
| AK002098.1 | 1 | 1690 | ||
| BQ184804.1 | 1 | 317 | ||
| KU159426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602970 | Chr.6:41070039 - 41258521 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map