Assay Details
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGM3 | NM_001199917.1 | NP_001186846.1 | ||
| NM_001199918.1 | NP_001186847.1 | |||
| NM_001199919.1 | NP_001186848.1 | |||
| NM_015599.2 | NP_056414.1 | |||
| XM_017010935.1 | XP_016866424.1 | |||
| XM_017010936.1 | XP_016866425.1 | |||
| XM_017010937.1 | XP_016866426.1 | |||
| XM_017010938.1 | XP_016866427.1 | |||
| AB032081.1 | BAB00613.1 | |||
| AF102265.1 | AAC72409.1 | |||
| AF180371.1 | AAD55097.1 | |||
| AK001365.1 | ||||
| AK023432.1 | ||||
| AK225363.1 | ||||
| AK301867.1 | ||||
| AK314512.1 | ||||
| AL117443.1 | CAB55928.1 | |||
| BC001258.1 | AAH01258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map