Assay Details
Target Gene Details
Entrez Gene ID: | 54516 |
Gene Name: | mitochondrial translational release factor 1 like |
Gene Aliases: |
HMRF1L, MRF1L, mtRF1a |
Location: |
Chr.6:152987349-153007301 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTRF1L | XM_017010959.1 | 1 | 3627 | XP_016866448.1 |
More Information
Set Membership: |
|

Genomic Map