Assay Details
Target Gene Details
Entrez Gene ID: | 9258 |
Gene Name: | malignant fibrous histiocytoma amplified sequence 1 |
Gene Aliases: |
LRRC65, MASL1 |
Location: |
Chr.8:8783354-8893621 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFHAS1 | NM_004225.2 | NP_004216.2 | ||
| AB016816.1 | BAA74737.1 | |||
| AK027007.1 | ||||
| BC014226.2 | AAH14226.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610189 | Chr.8:8732927 - 8807430 on Build GRCh38 | Loss |
|
| nsv831228 | Chr.8:8750877 - 8933280 on Build GRCh38 | Loss |
|
| nsv1018828 | Chr.8:8752370 - 8933150 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map