Assay Details
Target Gene Details
Entrez Gene ID: | 2765 |
Gene Name: | glycosylphosphatidylinositol anchored molecule like |
Gene Aliases: |
LY6DL |
Location: |
Chr.8:142834801-142846846 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GML | NM_002066.2 | NP_002057.1 | ||
| BC126336.1 | ||||
| BC126338.1 | ||||
| D84290.1 | BAA12300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023021 | Chr.8:142717134 - 142851742 on Build GRCh38 | Loss |
|
| esv2661137 | Chr.8:142256294 - 142935448 on Build GRCh38 | Deletion |
|
| nsv612714 | Chr.8:142786489 - 142846817 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map