Assay Details
Target Gene Details
Entrez Gene ID: | 55246 |
Gene Name: | coiled-coil domain containing 25 |
Gene Aliases: |
- |
Location: |
Chr.8:27733316-27772656 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC25 | NM_001304529.1 | NP_001291458.1 | ||
| NM_001304530.1 | NP_001291459.1 | |||
| NM_001304532.1 | NP_001291461.1 | |||
| NM_018246.2 | NP_060716.2 | |||
| NR_130761.1 | ||||
| NR_130762.1 | ||||
| AK001715.1 | BAA91857.1 | |||
| AK027900.1 | BAB55442.1 | |||
| AK292875.1 | ||||
| AK294848.1 | ||||
| AK311437.1 | ||||
| AK316015.1 | ||||
| BC006239.2 | AAH06239.1 | |||
| BC032588.1 | ||||
| BC049197.1 | ||||
| BC054039.1 | AAH54039.1 | |||
| BC065295.1 | AAH65295.1 | |||
| BC110391.1 | ||||
| BG718771.1 | ||||
| BU602778.1 | ||||
| DA218015.1 |
Target Gene Details
Entrez Gene ID: | 157570 |
Gene Name: | establishment of sister chromatid cohesion N-acetyltransferase 2 |
Gene Aliases: |
2410004I17Rik, EFO2, RBS |
Location: |
Chr.8:27771949-27812404 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESCO2 | XM_011544421.2 | XP_011542723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017032 | Chr.8:27767011 - 27821659 on Build GRCh38 | Loss |
|
| nsv528584 | Chr.8:27771369 - 27772288 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map