Assay Details
Target Gene Details
Entrez Gene ID: | 114786 |
Gene Name: | XK related 4 |
Gene Aliases: |
XRG4 |
Location: |
Chr.8:55102457-55526155 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XKR4 | NM_052898.1 | NP_443130.1 | ||
| AB067476.1 | BAB67782.1 | |||
| AY534241.1 | AAT07090.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74533777] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map