Assay Details
Target Gene Details
Entrez Gene ID: | 10 |
Gene Name: | N-acetyltransferase 2 |
Gene Aliases: |
AAC2, NAT-2, PNAT |
Location: |
Chr.8:18386585-18401219 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT2 | NM_000015.2 | NP_000006.2 | ||
| XM_017012938.1 | XP_016868427.1 | |||
| AJ581144.1 | CAE46171.1 | |||
| BC015878.1 | AAH15878.1 | |||
| BC067218.1 | AAH67218.1 | |||
| D90040.1 | BAA14094.1 | |||
| D90042.1 | BAA14096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428196 | Chr.8:18219826 - 18542377 on Build GRCh38 | Gain |
|
| nsv610717 | Chr.8:18327655 - 18400806 on Build GRCh38 | Gain |
|
| dgv7125n100 | Chr.8:18325949 - 18404169 on Build GRCh38 | Gain |
|
| nsv831252 | Chr.8:18347493 - 18548287 on Build GRCh38 | Gain |
|
| nsv1026366 | Chr.8:18071966 - 18557180 on Build GRCh38 | Gain |
|
| dgv12046n54 | Chr.8:18223142 - 18409264 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map