Assay Details
Target Gene Details
Entrez Gene ID: | 157506 |
Gene Name: | retinol dehydrogenase 10 (all-trans) |
Gene Aliases: |
SDR16C4 |
Location: |
Chr.8:73294602-73325285 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDH10 | NM_172037.4 | NP_742034.1 | ||
| AF456765.1 | AAN64747.1 | |||
| AY358270.1 | AAQ88637.1 | |||
| BC067131.1 | AAH67131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2737154 | Chr.8:71825978 - 73469084 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map