Assay Details
Target Gene Details
Entrez Gene ID: | 3551 |
Gene Name: | inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta |
Gene Aliases: |
IKK-beta, IKK2, IKKB, IMD15, NFKBIKB |
Location: |
Chr.8:42271302-42332653 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831301 | Chr.8:42244080 - 42473666 on Build GRCh38 | Loss |
|
| nsv508508 | Chr.8:42256602 - 42355479 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map