Assay Details
Target Gene Details
Entrez Gene ID: | 10404 |
Gene Name: | carboxypeptidase Q |
Gene Aliases: |
LDP, PGCP |
Location: |
Chr.8:96645227-97143503 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPQ | NM_016134.3 | NP_057218.1 | ||
| XM_017012953.1 | XP_016868442.1 | |||
| XM_017012954.1 | XP_016868443.1 | |||
| XM_017012955.1 | XP_016868444.1 | |||
| AF107834.1 | AAD43214.1 | |||
| AF119386.1 | AAD31418.1 | |||
| AK075132.1 | BAC11423.1 | |||
| AK296013.1 | ||||
| AK304008.1 | ||||
| AK315447.1 | ||||
| BC020689.1 | AAH20689.1 |
Target Gene Details
Entrez Gene ID: | 101927066 |
Gene Name: | uncharacterized LOC101927066 |
Gene Aliases: |
- |
Location: |
Chr.8:96951864-97446497 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927066 | NR_125390.1 | |||
| AK091342.1 |
More Information
Set Membership: |
|

Genomic Map