Assay Details
Target Gene Details
Entrez Gene ID: | 28998 |
Gene Name: | mitochondrial ribosomal protein L13 |
Gene Aliases: |
L13, L13A, L13mt, RPL13, RPML13 |
Location: |
Chr.8:120395844-120445407 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL13 | NM_014078.5 | NP_054797.2 | ||
| AB049640.1 | BAB40845.1 | |||
| AF112214.1 | AAF17202.1 | |||
| AK307322.1 | ||||
| AK311924.1 | ||||
| BC009190.2 | AAH09190.1 | |||
| BC021744.2 | AAH21744.1 | |||
| CN362173.1 |
More Information
Set Membership: |
|

Genomic Map