Assay Details
Target Gene Details
Entrez Gene ID: | 23414 |
Gene Name: | zinc finger protein, FOG family member 2 |
Gene Aliases: |
DIH3, FOG2, SRXY9, ZC2HC11B, ZNF89B, hFOG-2 |
Location: |
Chr.8:105318859-105804539 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFPM2 | NM_012082.3 | NP_036214.2 | ||
| XM_011516948.2 | XP_011515250.1 | |||
| AF119334.1 | AAD49558.1 | |||
| BC109222.1 | AAI09223.1 | |||
| BC109223.1 | AAI09224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521187 | Chr.8:105311508 - 105352623 on Build GRCh38 | Loss |
|
| nsv611837 | Chr.8:105298349 - 105359952 on Build GRCh38 | Loss |
|
| nsv1034972 | Chr.8:105294788 - 105357360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map