Assay Details
Target Gene Details
Entrez Gene ID: | 28957 |
Gene Name: | mitochondrial ribosomal protein S28 |
Gene Aliases: |
HSPC007, MRP-S28, MRP-S35, MRPS35 |
Location: |
Chr.8:79918860-80030271 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS28 | NM_014018.2 | NP_054737.1 | ||
| AF070663.1 | AAD20969.1 | |||
| AK026506.1 | ||||
| AK223069.1 | BAD96789.1 | |||
| AK315739.1 | ||||
| BC010150.1 | AAH10150.1 | |||
| KU178612.1 | ||||
| KU178613.1 | ||||
| KU178614.1 |
More Information
Set Membership: |
|

Genomic Map