Assay Details
Target Gene Details
Entrez Gene ID: | 100507316 |
Gene Name: | MYC-induced long noncoding RNA |
Gene Aliases: |
LINC01604 |
Location: |
Chr.8:143280155-143281700 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MINCR | NR_120682.1 | |||
| NR_120683.1 | ||||
| AA284855.1 | 1 | 238 | ||
| AA992445.1 | ||||
| AJ346444.1 | ||||
| AW975318.1 | ||||
| BM987389.1 | ||||
| HY339014.1 | ||||
| HY368981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950010 | Chr.8:143243031 - 143345330 on Build GRCh38 | Deletion |
|
| nsv1016726 | Chr.8:143251666 - 143317866 on Build GRCh38 | Loss |
|

Genomic Map