Assay Details
Target Gene Details
Entrez Gene ID: | 157724 |
Gene Name: | solute carrier family 7 member 13 |
Gene Aliases: |
AGT-1, AGT1, XAT2 |
Location: |
Chr.8:86213909-86230380 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A13 | NM_138817.2 | NP_620172.2 | ||
| XM_011516867.1 | XP_011515169.1 | |||
| AJ417661.1 | CAD10393.1 | |||
| AK055815.1 | BAB71021.1 | |||
| BC125165.1 | ||||
| BC125166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761445 | Chr.8:86173241 - 86322968 on Build GRCh38 | Gain |
|
| esv2762757 | Chr.8:86216014 - 86259547 on Build GRCh38 | Gain |
|
| nsv519507 | Chr.8:86174966 - 86321739 on Build GRCh38 | Gain+Loss |
|
| nsv6290 | Chr.8:86163511 - 86217579 on Build GRCh38 | Deletion |
|
| nsv428204 | Chr.8:85493249 - 86299979 on Build GRCh38 | Gain |
|
| dgv7263n100 | Chr.8:86149208 - 86325031 on Build GRCh38 | Gain |
|
| esv3617906 | Chr.8:86189102 - 86323318 on Build GRCh38 | Gain |
|
| dgv12228n54 | Chr.8:86174821 - 86321739 on Build GRCh38 | Gain |
|
| nsv611674 | Chr.8:86143943 - 86303691 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map