Assay Details
Target Gene Details
Entrez Gene ID: | 92421 |
Gene Name: | charged multivesicular body protein 4C |
Gene Aliases: |
SNF7-3, Shax3, VPS32C |
Location: |
Chr.8:81732453-81759515 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP4C | NM_152284.3 | NP_689497.1 | ||
| AB120734.1 | BAC87888.1 | |||
| AK000049.1 | ||||
| AK314873.1 | ||||
| AY329086.1 | AAQ91195.1 | |||
| BC014321.1 | AAH14321.1 | |||
| BX492886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv465721 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain |
|
| nsv831369 | Chr.8:81605833 - 81823586 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74544416] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map