Assay Details
Target Gene Details
Entrez Gene ID: | 767 |
Gene Name: | carbonic anhydrase 8 |
Gene Aliases: |
CA-RP, CA-VIII, CALS, CAMRQ3, CARP |
Location: |
Chr.8:60185420-60281423 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA8 | NM_001321839.1 | NP_001308768.1 | ||
| NM_004056.5 | NP_004047.3 | |||
| NR_135821.1 | ||||
| XM_017013818.1 | XP_016869307.1 | |||
| AK090655.1 | ||||
| AK293999.1 | ||||
| AY075022.1 | AAL78170.1 | |||
| BC069744.1 | AAH69744.1 | |||
| BC069794.1 | AAH69794.1 | |||
| BC108929.2 | AAI08930.1 | |||
| L04656.1 | AAA35653.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831333 | Chr.8:60190303 - 60362563 on Build GRCh38 | Gain |
|
| nsv831332 | Chr.8:60073041 - 60245171 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73683372] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map