Assay Details
Target Gene Details
Entrez Gene ID: | 81501 |
Gene Name: | dendrocyte expressed seven transmembrane protein |
Gene Aliases: |
FIND, TM7SF4, hDC-STAMP |
Location: |
Chr.8:104339087-104356874 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCSTAMP | XM_011517321.1 | XP_011515623.1 | ||
| XM_011517324.1 | XP_011515626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032775 | Chr.8:104299136 - 104397744 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map