Assay Details
Target Gene Details
Entrez Gene ID: | 105375843 |
Gene Name: | uncharacterized LOC105375843 |
Gene Aliases: |
- |
Location: |
Chr.8:55517187-55520978 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375843 | NR_134323.1 | |||
| AA412543.1 | ||||
| CR738863.1 |
Target Gene Details
Entrez Gene ID: | 114786 |
Gene Name: | XK related 4 |
Gene Aliases: |
XRG4 |
Location: |
Chr.8:55102457-55526155 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XKR4 | NM_052898.1 | NP_443130.1 | ||
| AB067476.1 | BAB67782.1 | |||
| AY534241.1 | AAT07090.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8348 | Chr.8:55168964 - 55539363 on Build GRCh38 | Loss |
|

Genomic Map