Assay Details
Target Gene Details
Entrez Gene ID: | 7201 |
Gene Name: | thyrotropin releasing hormone receptor |
Gene Aliases: |
TRH-R |
Location: |
Chr.8:109085745-109119785 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRHR | NM_003301.5 | NP_003292.1 | ||
| XM_011517263.2 | XP_011515565.1 | |||
| AY493373.1 | AAR84356.1 | |||
| BC105045.1 | AAI05046.1 | |||
| BC113360.1 | ||||
| D16845.1 | BAA04120.1 | |||
| X72089.1 | CAA50979.1 | |||
| X75071.1 | CAA52965.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73311314,rs75528129] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map